Learn More
(5-Methylthien-2-yl)methylamine hydrochloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 163.67 |
Percent Purity | 97% |
Chemical Name or Material | (5-methylthien-2-yl)methylamine hydrochloride |