Läs mer
5-Methyluridine-1',2',3',4',5'-13C5, TRC
Produktkod. 30467408
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30467408 5 mg
30471825 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30467408 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3381985MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 159496-17-0
Molekylformel 13C5C5H14N2O6
Molekylvikt (g/mol) 263.19
Synonym Ribothymidine-1’,2’,3’,4’,5’-13C
LEDER O[13C@@H]1[13C@H](O)[13C@@H]([13CH2]O)O[13C@H]1N2C=C(C)C(NC2=O)=O
Specifikationer
Kemiskt namn eller material 5-Methyluridine-1',2',3',4',5'-13C5
Kvantitet 5 mg
InChI formel InChI=1S/C10H14N2O6/c1-4-2-12(10(17)11-8(4)16)9-7(15)6(14)5(3-13)18-9/h2,5-7,9,13-15H,3H2,1H3,(H,11,16,17)/t5-,6-,7-,9-/m1/s1/i3+1,5+1,6+1,7+1,9+1