missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Nitro-1-benzothiophene-2-carbonyl chloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 241.65 |
Percent Purity | 95% |
Chemical Name or Material | 5-Nitro-1-benzothiophene-2-carbonyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'