missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Nitro-2-(4-methylphenyl)benzimidazole, 95%

Brand:  Alfa Aesar™ H64316.03

Additional Details : CAS Number : 1571-90-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15491459

  • 3114.20 SEK / 1g


Estimated Shipment date
from Supplier 06-08-2018
Add to basket

Specifications

Chemical Identifiers

5-Nitro-2-(4-methylphenyl)benzimidazole
1571-90-0
MFCD00430513
5-Nitro-2-(p-tolyl)benzimidazole

Specifications

5-Nitro-2-(4-methylphenyl)benzimidazole
1g
Solid
C14H11N3O2
5-Nitro-2-(p-tolyl)benzimidazole
CC1=CC=C(C=C1)C2=NC3=C(N2)C=C(C=C3)[N+](=O)[O-]
376349
253.26
1571-90-0
95%
MFCD00430513
AAMPIRQLXWIYCV-UHFFFAOYSA-N
253.261
Videos
SDS