Läs mer
5-Nitro-6-methyluracil, 99%, Thermo Scientific™
Produktkod. p-7037229
Change view
Kvantitet:
250 mg
1 g
5 g
Förpackningsstorlek:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
15472308 250 mg 250mg
15452308 1 g 1g
15462308 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 15472308 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H55913.MD
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
5-Nitro-6-methyluracil, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in DMF, DMSO, hot methanol and hot water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 16632-21-6
Molekylformel C5H5N3O4
Molekylvikt (g/mol) 171.11
MDL-nummer MFCD00051201
InChI-nyckel LIVYMRJSNFHYEN-UHFFFAOYSA-N
Synonym 5-nitro-6-methyluracil, 6-methyl-5-nitrouracil, 6-methyl-5-nitropyrimidine-2,4 1h,3h-dione, 2,4-dihydroxy-6-methyl-5-nitropyrimidine, 6-metyl-5-nitrouracil, 6-methyl-5-nitropyrimidine-2,4-diol, uracil, 6-methyl-5-nitro, 2,4 1h,3h-pyrimidinedione, 6-methyl-5-nitro, 4-methyl-5-nitrouracil, 2,4-dihydroxy-6-methyl-5-*nitropyrimidine
PubChem CID 85523
IUPAC-namn 6-methyl-5-nitro-1H-pyrimidine-2,4-dione
LEDER CC1=C(C(=O)NC(=O)N1)[N+]([O-])=O
Specifikationer
Kemiskt namn eller material 5-Nitro-6-methyluracil
Kvantitet 250 mg
Löslighetsinformation Soluble in DMF,DMSO,hot methanol and hot water.
Formel vikt 171.11
Procent renhet 99%