missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 5-Nitroindole-3-carboxaldehyde, 95%
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Quantity | 250 mg |
Beilstein | 1530696 |
Formula Weight | 190.16 |
Percent Purity | 95% |
Physical Form | Solid |
Chemical Name or Material | 5-Nitroindole-3-carboxaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'