Läs mer
5-Nitroisoindoline Hydrochloride Salt, TRC
Produktkod. 30471959
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30471959 250 mg
1 options
Produktkod. 30471959 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN496370250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5-Nitroisoindoline Hydrochloride Salt
CAS 400727-69-7
Molekylformel C8H9ClN2O2
Kvantitet 250 mg
Synonym 2,3-Dihydro-5-nitro-1H-isoindole Hydrochloride Salt, 5-Nitro-2,3-dihydro-1H-isoindole Hydrochloride Salt, 1H-Isoindole','2,3-dihydro-5-nitro-','monohydrochloride (9CI), 1H-Isoindole 2,3-dihydro-5-nitro-','hydrochloride (1:1), 5-Nitroisoindoline hydrochloride
InChI formel InChI=1S/C8H8N2O2.ClH/c11-10(12)8-2-1-6-4-9-5-7(6)3-8;/h1-3,9H,4-5H2;1H
LEDER C1=C2CNCC2=CC(=C1)[N+](=O)[O-].Cl
IUPAC-namn 5-nitroisoindoline Hydrochloride Salt
Molekylvikt (g/mol) 200.62
Formel vikt 200.04