Läs mer
5-Nitroso-quinolin-8-ol, TRC
Produktkod. 30136497
Change view
Kvantitet:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136497 100 mg
30134503 50 mg
30004012 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136497 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN546580100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3565-26-2
Molekylformel C9H6N2O2
Molekylvikt (g/mol) 174.16
Synonym 5-Nitroso-8-quinolinol, 5-Nitroso-8-hydroxyquinoline, 5-Nitrosooxine, Hydron III, NSC 3852, 8-Hydroxy-5-nitrosoquinoline
IUPAC-namn 5-nitrosoquinolin-8-ol
LEDER C1=CC2=C(C(=CC=C2N=O)O)N=C1
Specifikationer
Kemiskt namn eller material 5-Nitroso-quinolin-8-ol
Kvantitet 100 mg
InChI formel InChI=1S/C9H6N2O2/c12-8-4-3-7(11-13)6-2-1-5-10-9(6)8/h1-5,12H
Formel vikt 174.0429