missing translation for 'onlineSavingsMsg'
Learn More

5-Norbornene-2-carboxylic acid, 98%, mixture of isomers, ACROS Organics™

Brand:  Acros Organics 453301000

Additional Details : CAS Number : 120-74-1 Weight : 0.10000kg Transport : UN number : 3265 Chem class : 8 Pack group : II


Quantity 100g

 View more versions of this product

Product Code. 15209886

  • 1914.66 SEK / 100g


In stock, ready to ship: 2
Add to basket


Specifications

Chemical Identifiers

120-74-1
138.16
5-norbornene-2-carboxylic acid, bicyclo 2.2.1 hept-5-ene-2-carboxylic acid, norbornenecarboxylic acid, exo-5-norbornene-2-carboxylic acid, 5-norbornene-2-carboxylic acid, exo, 5-norbornene-2-carboxylic acid 8ci, bicyclo 2.2.1 hept-2-ene-5-carboxylic acid, bicyclo 2.2.1-5-heptene-2-carboxylic acid, norborn-5-ene-2-carboxylic acid
bicyclo[2.2.1]hept-2-ene-5-carboxylic acid
C8H10O2
FYGUSUBEMUKACF-UHFFFAOYSA-N
78949
C1C2CC(C1C=C2)C(=O)O

Specifications

5-Norbornene-2-carboxylic acid
>110°C
1.129
100g
97.5% min. (total of isomers) (GC)
5-norbornene-2-carboxylic acid, bicyclo 2.2.1 hept-5-ene-2-carboxylic acid, norbornenecarboxylic acid, exo-5-norbornene-2-carboxylic acid, 5-norbornene-2-carboxylic acid, exo, 5-norbornene-2-carboxylic acid 8ci, bicyclo 2.2.1 hept-2-ene-5-carboxylic acid, bicyclo 2.2.1-5-heptene-2-carboxylic acid, norborn-5-ene-2-carboxylic acid
C1C2CC(C1C=C2)C(=O)O
138.16
138.16
1.1290g/mL
Authentic
136.0°C to 138.0°C (14.0mmHg)
120-74-1
C8H10O2
FYGUSUBEMUKACF-UHFFFAOYSA-N
bicyclo[2.2.1]hept-2-ene-5-carboxylic acid
78949
98%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 5-Norbornene-2-carboxylic acid

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Documents