Learn More
5-Norbornene-2-carboxylic acid, 98%, mixture of isomers, Thermo Scientific Chemicals
Catalog Number 15289876
Click to view available options
Quantity:
5 g
100 g
120-74-1
C8H10O2
138.16
FYGUSUBEMUKACF-UHFFFAOYSA-N
5-norbornene-2-carboxylic acid, bicyclo 2.2.1 hept-5-ene-2-carboxylic acid, norbornenecarboxylic acid, exo-5-norbornene-2-carboxylic acid, 5-norbornene-2-carboxylic acid, exo, 5-norbornene-2-carboxylic acid 8ci, bicyclo 2.2.1 hept-2-ene-5-carboxylic acid, bicyclo 2.2.1-5-heptene-2-carboxylic acid, norborn-5-ene-2-carboxylic acid
78949
bicyclo[2.2.1]hept-2-ene-5-carboxylic acid
C1C2CC(C1C=C2)C(=O)O
120-74-1
C8H10O2
138.16
FYGUSUBEMUKACF-UHFFFAOYSA-N
5-norbornene-2-carboxylic acid, bicyclo 2.2.1 hept-5-ene-2-carboxylic acid, norbornenecarboxylic acid, exo-5-norbornene-2-carboxylic acid, 5-norbornene-2-carboxylic acid, exo, 5-norbornene-2-carboxylic acid 8ci, bicyclo 2.2.1 hept-2-ene-5-carboxylic acid, bicyclo 2.2.1-5-heptene-2-carboxylic acid, norborn-5-ene-2-carboxylic acid
78949
bicyclo[2.2.1]hept-2-ene-5-carboxylic acid
C1C2CC(C1C=C2)C(=O)O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Density 1.1290g/mL
Boiling Point 136.0°C to 138.0°C (14.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (total of isomers) (GC)
Quantity 5 g
Specific Gravity 1.129
Formula Weight 138.16
Percent Purity 98%
Chemical Name or Material 5-Norbornene-2-carboxylic acid
compliance-icons
Product Identifier
  • 5-Norbornene-2-carboxylic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.