missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Norbornene-2-carboxylic acid, 98%, mixture of isomers, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 1.1290g/mL |
Boiling Point | 136.0°C to 138.0°C (14.0 mmHg) |
Flash Point | >110°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (total of isomers) (GC) |
Quantity | 5 g |
Specific Gravity | 1.129 |
Formula Weight | 138.16 |
Percent Purity | 98% |
Chemical Name or Material | 5-Norbornene-2-carboxylic acid |
Safety and Handling
Product Identifier
- 5-Norbornene-2-carboxylic acid
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'