Läs mer
5-O-Desmethyl Donepezil, TRC
Produktkod. 30138711
Change view
Kvantitet:
2.5 mg
5 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30138711 25 mg -
30010728 5 mg -
30008775 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138711 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29201025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 120013-57-2
Molekylformel C23 H27 N O3
Molekylvikt (g/mol) 365.47
Synonym 1H-Inden-1-one, 2,3-dihydro-5-hydroxy-6-methoxy-2-[[1-(phenylmethyl)-4-piperidinyl]methyl]-, 2,3-Dihydro-5-hydroxy-6-methoxy-2-[[1-(phenylmethyl)-4-piperidinyl]methyl]-1H-inden-1-one, 1-Benzyl-4-[(5-hydroxy-6-methoxy-1-oxo-indan-2-yl)methyl]piperidine, 5-O-Desmethyl donepezil
IUPAC-namn 2-[(1-benzylpiperidin-4-yl)methyl]-5-hydroxy-6-methoxy-2,3-dihydroinden-1-one
LEDER COc1cc2C(=O)C(CC3CCN(Cc4ccccc4)CC3)Cc2cc1O
Specifikationer
Kemiskt namn eller material 5-O-Desmethyl Donepezil
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C23H27NO3/c1-27-22-14-20-18(13-21(22)25)12-19(23(20)26)11-16-7-9-24(10-8-16)15-17-5-3-2-4-6-17/h2-6,13-14,16,19,25H,7-12,15H2,1H3
Formel vikt 365.1991
Procent renhet >95%