Läs mer
5-Oxo-1,3-dioxolane-4,4-diacetic Acid, TRC
Produktkod. 30423296
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30423296 10 g
30412968 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30423296 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO85550010G
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 144-16-1
Molekylformel C7H8O7
Molekylvikt (g/mol) 204.13
Synonym beta-(Hydroxymethoxy)-tricarballylic acid gamma-Lactone, Anhydromethylenecitric Acid, Methylenecitric Acid, NSC 68614
IUPAC-namn 2-[4-(carboxymethyl)-5-oxo-1,3-dioxolan-4-yl]acetic acid
LEDER OC(=O)CC1(CC(=O)O)OCOC1=O
Specifikationer
Kemiskt namn eller material 5-Oxo-1,3-dioxolane-4,4-diacetic Acid
Kvantitet 10 g
InChI formel InChI=1S/C7H8O7/c8-4(9)1-7(2-5(10)11)6(12)13-3-14-7/h1-3H2,(H,8,9)(H,10,11)
Formel vikt 204.027