missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Oxo-5-[phenyl[1-(2-phenylethyl)-4-piperidinyl]amino]pentanoic Acid, TRC
Specifications
Specifications
| Kemiskt namn eller material | Valeryl fentanyl carboxy metabolite |
| CAS | 526191-19-5 |
| Molekylformel | C24 H30 N2 O3 |
| Kvantitet | 100 mg |
| Synonym | Pentanoyl fentanyl carboxy metabolite, Pentanyl fentanyl carboxy metabolite |
| InChI formel | InChI=1S/C24H30N2O3/c27-23(12-7-13-24(28)29)26(21-10-5-2-6-11-21)22-15-18-25(19-16-22)17-14-20-8-3-1-4-9-20/h1-6,8-11,22H,7,12-19H2,(H,28,29) |
| LEDER | OC(=O)CCCC(=O)N(C1CCN(CCc2ccccc2)CC1)c3ccccc3 |
| IUPAC-namn | 5-oxo-5-(N-[1-(2-phenylethyl)piperidin-4-yl]anilino)pentanoic acid |
| Molekylvikt (g/mol) | 394.507 |
| Formel vikt | 394.226 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?