Läs mer
5-Oxohexanoic Acid Ethyl Ester, TRC
Produktkod. 30411594
Change view
Kvantitet:
25 g
Produktkod. Kvantitet
30411594 25 g
1 options
Produktkod. 30411594 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO85686525G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5-Oxohexanoic Acid Ethyl Ester
CAS 13984-57-1
Molekylformel C8H14O3
Kvantitet 25 g
Synonym 4-Acetylbutyric Acid Ethyl Ester, Ethyl 5-oxocaproate, Ethyl 5-oxohexanoate
InChI formel InChI=1S/C8H14O3/c1-3-11-8(10)6-4-5-7(2)9/h3-6H2,1-2H3
LEDER CCOC(=O)CCCC(=O)C
IUPAC-namn ethyl 5-oxohexanoate
Molekylvikt (g/mol) 158.19
Formel vikt 158.0943