missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Tocris Bioscience™ 5-Ph-IAA

Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
50 mg
Beskrivning
5-Ph-IAA is a selective and potent "Bump & Hole" TAG Degrader (DC50,6h = 17.0 nM, T1/2 = 62.3 min), for use within an auxin-inducible degron (AID2) system. 5-Ph-IAA is an analog (the “Bump”) of natural auxin that induces rapid degradation of a target protein tagged with a single-point mutant OsTIR1F74G F-box protein (the "Hole" or mini AID). The induced degradation is reversible 3 h after washout with fresh medium. 5-Ph-IAA shows rapid depletion of mAID-tagged nuclear and cytoplasmic proteins in proliferating and non-proliferating mammalian cells with no detectable basal degradation, lower cytotoxicity level, increased potency and faster degradation of the reporter in comparison with the conventional AID system. The AID2 system also works in budding yeast and fission yeast, and it can control protein expression in multiple mouse organs in vivo.
To enquire about custom knock-in cell lines for the AID2 platform, please email TPD@bio-techne.com
Plasmid vectors for the AAV transfection and CRISPR-mediated knock-in of OsTIR1F74G and mAID tag are available from Addgene .
Specifikationer
Specifikationer
| Kemiskt namn eller material | 5-Phenyl-1 H-indole-3-acetic acid |
| CAS | 168649-23-8 |
| Kvantitet | 50 mg |
| Mål | Active Degraders |
| Molekylformel | C16H13NO2 |
| Renhet | 0.98 |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering