Läs mer
5-Phenylbenzene-1,3-diol, TRC
Produktkod. 30424574
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30424574 50 mg
30419931 10 mg
30414247 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30424574 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP32232350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7028-41-3
Molekylformel C12H10O2
Molekylvikt (g/mol) 186.21
IUPAC-namn 5-phenylbenzene-1,3-diol
LEDER Oc1cc(O)cc(c1)c2ccccc2
Specifikationer
Kemiskt namn eller material 5-Phenylbenzene-1,3-diol
Kvantitet 50 mg
InChI formel InChI=1S/C12H10O2/c13-11-6-10(7-12(14)8-11)9-4-2-1-3-5-9/h1-8,13-14H
Formel vikt 186.0681