Läs mer
5-Propylhydantoin, TRC
Produktkod. 30426398
Change view
Kvantitet:
250 mg
500 mg
2.5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30426398 500 mg -
30416957 2.5 g 2.5g
30416755 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30426398 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP837115500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 18227-41-3
Molekylformel C6H10N2O2
Molekylvikt (g/mol) 142.15
IUPAC-namn 5-propylimidazolidine-2,4-dione
LEDER CCCC1NC(=O)NC1=O
Specifikationer
Kemiskt namn eller material 5-Propylhydantoin
Kvantitet 500 mg
InChI formel InChI=1S/C6H10N2O2/c1-2-3-4-5(9)8-6(10)7-4/h4H,2-3H2,1H3,(H2,7,8,9,10)
Formel vikt 142.0742