Läs mer
[5-(Pyridin-2-yl)-1H-imidazol-2-yl]methanamine Trihydrochloride, TRC
Produktkod. 30420651
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30420651 10 mg
30423059 100 mg
30425896 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30420651 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP99476010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2060007-34-1
Molekylformel C9H10N4 • 3HCl
Molekylvikt (g/mol) 283.59
IUPAC-namn (5-(pyridin-2-yl)-1H-imidazol-2-yl)methanamine trihydrochloride
LEDER NCC(N1)=NC=C1C2=CC=CC=N2.Cl.Cl.Cl
Specifications
Kemiskt namn eller material [5-(Pyridin-2-yl)-1H-imidazol-2-yl]methanamine Trihydrochloride
Kvantitet 10 mg
InChI formel InChI=1S/C9H10N4.3ClH/c10-5-9-12-6-8(13-9)7-3-1-2-4-11-7;;;/h1-4,6H,5,10H2,(H,12,13);3*1H
Formel vikt 282.03