Läs mer
5-Pyridin-4-yl-1H-indazole, TRC
Produktkod. 30413467
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30413467 5 mg
30419512 10 mg
30425924 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30413467 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP9975985MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 885272-55-9
Molekylformel C12H9N3
Molekylvikt (g/mol) 195.22
IUPAC-namn 5-pyridin-4-yl-2H-indazole
LEDER c1cc(ccn1)c2ccc3n[nH]cc3c2
Specifikationer
Kemiskt namn eller material 5-Pyridin-4-yl-1H-indazole
Kvantitet 5 mg
InChI formel InChI=1S/C12H9N3/c1-2-12-11(8-14-15-12)7-10(1)9-3-5-13-6-4-9/h1-8H,(H,14,15)
Formel vikt 195.0796