missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Pyrimidineacetic acid, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (HPLC) |
Packaging | Glass Bottle |
Molecular Formula | C6H6N2O2 |
Quantity | 1 g |
Synonym | alitretinoin, 9-cis-retinoic acid, panretin, 9-cis-tretinoin, panrexin, panretyn, panretin gel, toctino, 9-cis retinoic acid, 9 z-retinoic acid |
SMILES | OC(=O)CC1=CN=CN=C1 |
IUPAC Name | (2E,4E,6Z,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid |
PubChem CID | 449171 |
Formula Weight | 138.13 |
Show More |
Safety and Handling
Product Identifier
- 5-Pyrimidineacetic acid, 95%
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'