Learn More
5-Pyrimidineacetic acid, 95%, Thermo Scientific™
Catalog Number p-7077471
Click to view available options
Quantity:
250 mg
1 g
5267-07-2
138.13
HEEUMCHKVXTQIP-UHFFFAOYSA-N
5267-07-2
138.13
HEEUMCHKVXTQIP-UHFFFAOYSA-N
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Molecular Formula C6H6N2O2
Quantity 250 mg
Synonym 5-pyrimidineacetic acid, 2-pyrimidin-5-yl acetic acid, 5-pyrimidine acetic acid, pyrimidin-5-ylacetic acid, 5-pyrimidineacetate, pubchem13279, pyrimidine-5-acetic acid, pyrimidin-5-yl-acetic acid, pyrimidin-5-yl acetic acid, 5-pyrimidineacetic acid 7ci,8ci,9ci
SMILES C1=C(C=NC=N1)CC(=O)O
IUPAC Name 2-pyrimidin-5-ylacetic acid
PubChem CID 21471675
Formula Weight 138.13
Percent Purity 95%
Chemical Name or Material 5-Pyrimidineacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • 5-Pyrimidineacetic acid, 95%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.