missing translation for 'onlineSavingsMsg'
Learn More

5-Pyrimidineacetic acid, 95%, Acros Organics™

Brand:  Acros Organics 454282500

Additional Details : CAS Number : 5267-07-2 Weight : 0.00025kg


Quantity 250mg

 View more versions of this product

Product Code. 15244338

  • 1796.59 SEK / 250mg


Estimated Shipment date
from Supplier 06-03-2019
Add to basket


Specifications

Chemical Identifiers

5267-07-2
138.13
5-pyrimidineacetic acid, 2-pyrimidin-5-yl acetic acid, 5-pyrimidine acetic acid, pyrimidin-5-ylacetic acid, 5-pyrimidineacetate, pubchem13279, pyrimidine-5-acetic acid, pyrimidin-5-yl-acetic acid, pyrimidin-5-yl acetic acid, 5-pyrimidineacetic acid 7ci,8ci,9ci
2-pyrimidin-5-ylacetic acid
C6H6N2O2
HEEUMCHKVXTQIP-UHFFFAOYSA-N
21471675
C1=C(C=NC=N1)CC(=O)O

Specifications

5-Pyrimidineacetic acid
250mg
C6H6N2O2
HEEUMCHKVXTQIP-UHFFFAOYSA-N
2-pyrimidin-5-ylacetic acid
21471675
≥94.0% (HPLC)
Authentic
5267-07-2
5-pyrimidineacetic acid, 2-pyrimidin-5-yl acetic acid, 5-pyrimidine acetic acid, pyrimidin-5-ylacetic acid, 5-pyrimidineacetate, pubchem13279, pyrimidine-5-acetic acid, pyrimidin-5-yl-acetic acid, pyrimidin-5-yl acetic acid, 5-pyrimidineacetic acid 7ci,8ci,9ci
C1=C(C=NC=N1)CC(=O)O
138.13
138.13
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 5-Pyrimidineacetic acid, 95%

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.