missing translation for 'onlineSavingsMsg'
Läs mer

5-Trifluoromethyl-2H-pyridazine-3-one, TRC

Produktkod. 30450513
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30450513 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30450513 Supplier Toronto Research Chemicals Supplier No. TRCT791350250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 5-Trifluoromethyl-2H-pyridazine-2-one
CAS 244268-34-6
Molekylformel C5 H3 F3 N2 O
Kvantitet 250 mg
InChI formel InChI=1S/C5H3F3N2O/c6-5(7,8)3-1-4(11)10-9-2-3/h1-2H,(H,10,11)
LEDER FC(F)(F)C1=CC(=O)NN=C1
IUPAC-namn 4-(trifluoromethyl)-1H-pyridazin-6-one
Molekylvikt (g/mol) 164.09
Formel vikt 164.0197
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.