Läs mer
5-(Trifluoromethyl)indolin-2-one, TRC
Produktkod. 30450498
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30450498 100 mg
30453960 50 mg
30451145 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30450498 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT899643100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 71293-62-4
Molekylformel C9H6NOF3
Molekylvikt (g/mol) 201.15
Synonym 1,3-Dihydro-5-(trifluoromethyl)-2H-indol-2-one (ACI), 5-(Trifluoromethyl)-1,3-dihydroindol-2-one, 5-(Trifluoromethyl)-2-oxindole, 5-(Trifluoromethyl)indolin-2-one, 5-(Trifluoromethyl)oxindole
IUPAC-namn 5-(trifluoromethyl)indolin-2-one
LEDER O=C1NC2=C(C=C(C(F)(F)F)C=C2)C1
Specifikationer
Kemiskt namn eller material 5-(Trifluoromethyl)indolin-2-one
Kvantitet 100 mg
InChI formel InChI=1S/C9H6F3NO/c10-9(11,12)6-1-2-7-5(3-6)4-8(14)13-7/h1-3H,4H2,(H,13,14)
Formel vikt 201.04