Läs mer
5-(Trimethylstannyl)-2'-deoxyuridine, TRC
Produktkod. 30450655
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30450655 25 mg
30453517 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30450655 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT79720025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 146629-34-7
Molekylformel C12 H20 N2 O5 Sn
Molekylvikt (g/mol) 391.01
Synonym 2'-Deoxy-5-(trimethylstannyl)uridine
IUPAC-namn 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-trimethylstannylpyrimidine-2,4-dione
LEDER C[Sn](C)(C)C1=CN([C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)NC1=O
Specifikationer
Kemiskt namn eller material 5-(Trimethylstannyl)-2'-deoxyuridine
Kvantitet 25 mg
InChI formel InChI=1S/C9H11N2O5.3CH3.Sn/c12-4-6-5(13)3-8(16-6)11-2-1-7(14)10-9(11)15;;;;/h2,5-6,8,12-13H,3-4H2,(H,10,14,15);3*1H3;/t5-,6+,8+;;;;/m0..../s1
Formel vikt 392.0394