missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
R&D Systems™ Methyl Green 1%
Klicka för att se tillgängliga alternativ
Kvantitet:
50 mL
Description
- Uncontrolled product according to WHMIS classification criteria
- There are no hazardous ingredients as defined in the United States 29 CFR 1910.1200 (OSHA) or the Canadian Controlled Products Regulations (WHMIS) and therefore not subject to the labelling requirements of the aforementioned standards
Specifications
Specifications
| Kvantitet | 50 mL |
| Innehåll och lagring | Store the unopened product at room temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction