Learn More
Spectrum™ Labs Spectra/Por™ 1 6-8 kD MWCO Standard RC Dry Dialysis Kits
Product Code. 15350752
Change view
Click to view available options
Diameter (Metric):
6.4 mm
14.6 mm
20.4 mm
25.5 mm
32 mm
64 mm
76 mm
Length (Metric):
0.32 mL/cm vol./length
1.7 mL/cm
3.3 mL/cm vol./length
32 mL/cm vol./length
46 mL/cm vol./length
5 m
5.1 mL/cm vol./length
7.9 mL/cm vol./length
Quantity:
1 m Tubing/Kit
5 m Tubing/Kit
Each
Unit Size:
1m
5m
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Length (Metric) Quantity Unit Size
15350752 64 mm 32 mL/cm vol./length 5 m Tubing/Kit 5m
15370752 76 mm 46 mL/cm vol./length 5 m Tubing/Kit 5m
15380752 76 mm 46 mL/cm vol./length 1 m Tubing/Kit 1m
15330752 32 mm 7.9 mL/cm vol./length 1 m Tubing/Kit 1m
15360752 64 mm 32 mL/cm vol./length 1 m Tubing/Kit 1m
15320752 25.5 mm 5.1 mL/cm vol./length 1 m Tubing/Kit 1m
15300752 20.4 mm 3.3 mL/cm vol./length 1 m Tubing/Kit 1m
15310752 25.5 mm 5.1 mL/cm vol./length 5 m Tubing/Kit 5m
15380742 14.6 mm 1.7 mL/cm 1 m Tubing/Kit 1m
15370742 14.6 mm 1.7 mL/cm 5 m Tubing/Kit 5m
15350742 6.4 mm 0.32 mL/cm vol./length 5 m Tubing/Kit 5m
15360742 6.4 mm 0.32 mL/cm vol./length 1 m Tubing/Kit 1m
11511040 20.4 mm 5 m Each 5m
11521040 32 mm 5 m Each 5m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 15350752 Supplier Spectrum™ Supplier No. 132670T
  • ONLINE EXCLUSIVE PRICE
    2660.00 SEK
    2114.31 SEK / 5m
    Save 545.69 SEK
    20% Av
View all promotions
Beräknad leverans: 14-12-2026
Includes
  • 1m or 5m of Spectra/Por 1 dialysis tubing
  • 1 Standard Spectra/Por Closure (for buoyance)
  • 1 weighted Spectra/Por Closure (for orientation)
  • 2 adhesive tubing openers
With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated
  • For basic dialysis applications, or evaluating dialysis tubing for dialysis application
  • Packaged dry with glycerin (soak in water)
Specifications
Product Type Dialysis Trial Kit
MWCO 6 to 8 kDa
Format Tubing
Diameter (Metric) 64 mm
Length (Metric) 32 mL/cm vol./length
Quantity 5 m Tubing/Kit