missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ cGMP Recombinant Rabbit Monoclonal Antibody (RM467)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544557
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
This antibody reacts to the methoxy group of Polyethylene glycol (PEG). It is specific for methoxypoly(ethylene glycol).
Guanosine cyclic 3',5'-(hydrogen phosphate) is a guanine nucleotide containing one phosphate group which is esterified to the sugar moiety in both the 3'- and 5'-positions. Cyclic gmp is a cellular regulatory agent and has been described as a second messenger. Its levels increase in response to a variety of hormones, including acetylcholine, insulin, and oxytocin and it has been found to activate specific protein kinases. [National Center for Biotechnology Information. PubChem Compound Database; CID=24316].
Specifikationer
| cGMP | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide | |
| Rabbit | |
| Protein A | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
| ELISA | |
| RM467 | |
| Unconjugated | |
| C10H12N5O7P; Cyclic GMP | |
| KLH-conjugated cGMP. | |
| 50 μg | |
| Primary | |
| -20°C | |
| Liquid | |
| Chemical |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering