missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ 5424 Centrifuge with FA-45-24-11Model Rotor

Equipped for all modern molecular biology applications using Eppendorf and PCR tubes. Eppendorf™ 5424 Centrifuges are designed for separating liquid substance mixtures with different densities, and analyzing samples from the human body in in-vitro diagnostic applications.

Brand:  Eppendorf™ 5424000010

Additional Details : Weight : 17.10000kg

Product Code. 10584983

  • 24100.00 SEK / Each


Estimated Shipment
12-04-2019
Add to basket

Description

Description

  • Features state of the art cooling technology to ensure energy efficiency and high temperature accuracy for maximum sample protection
  • OptiBowl™ design for quiet operation, even without a rotor lid
  • Soft-touch one-finger lid closure for ergonomic operation
  • SOFT brake to protect sensitive samples
  • Full digital display with time and speed display
  • Separate short-spin key for quick centrifugation
  • Certified aerosol-tight rotor comes standard for safe centrifugation of hazardous samples
  • Available as a rotary knob version for fast parameter selection, or keypad version for easy cleaning
  • Temperature range from -10  to +40°C
  • Guaranteed temperature of 4°C at maximum rotational speed
  • FastTemp function for quick pre-cooling of the centrifuge, e.g. from ~ 21 to 4°C in only 8 minutes
  • ECO power shut-off engages after 8 hours of non-use to reduce energy consumption (40% over night) and extend compressor life (can be deactivated)
  • The patented compressor technology reduces vibrations and protects your samples
  • Built-in condensation drain to eliminate water accumulation in the rotor chamber
Specifications

Specifications

29.5 lb. (13.4kg)
20,238 x G
2 to 40°C
Keypad, with FA-45-24-11 Model Rotor
320mm
100 to 14680rpm
5424
24 x 2mL
<51dB(A)
Centrifuge
230V 50/60Hz
250W
30Sec to 9:59Hr
Videos
View accessory products