missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ 5424 Centrifuges without Rotor

For molecular and biological laboratories

Brand:  Eppendorf™ 5424000614

Additional Details : Weight : 16.20000kg

 View more versions of this product

Product Code. 10022962

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Choice of four different rotors (available separately)
  • Standard rotor can accommodate 1.5mL/2.0mL tubes
  • PCR strips and kits/spin columns can be centrifuged in the corresponding rotor
Specifications

Specifications

29.5 lb. (13.4kg)
20,238 x G
2 to 40°C
Dial
320mm
100 to 14680rpm
5424
24 x 2mL
<51dB(A)
Centrifuge
230V 50/60Hz
250W
30Sec to 9:59Hr
Videos
Documents
View accessory products