Läs mer
(5Alpha,10Alpha,17Beta)-5,10-Epoxy-17-hydroxy-17-(1-propyn-1-yl)-estr-9(11)-en-3-one Cyclic 1,2-Ethanediyl Acetal, TRC
Produktkod. 30376283
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30376283 5 mg
1 options
Produktkod. 30376283 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE5892105MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (5alpha,10alpha,17beta)-5,10-Epoxy-17-hydroxy-17-(1-propyn-1-yl)-estr-9(11)-en-3-one Cyclic 1,2-Ethanediyl Acetal
CAS 84371-57-3
Molekylformel C23H30O4
Kvantitet 5 mg
Synonym 5α,10α-Epoxy-3,3-(ethylenedioxy)-17β-hydroxy-17α-(1-propynyl)-9(11)-estrene, 5α,10α-Epoxy-17β-hydroxy-17-(1-propynyl)estr-9(11)-en-3-one Cyclic Ethylene Acetal
InChI formel InChI=1S/C23H30O4/c1-3-7-20(24)9-6-17-16-4-10-21-15-22(25-13-14-26-22)11-12-23(21,27-21)18(16)5-8-19(17,20)2/h5,16-17,24H,4,6,8-15H2,1-2H3/t16-,17-,19-,20-,21+,23+/m0/s1
LEDER CC#C[C@@]1(CC[C@@H]2[C@@]1(CC=C3[C@H]2CC[C@@]45[C@@]3(O4)CCC6(C5)OCCO6)C)O
Molekylvikt (g/mol) 370.48
Formel vikt 370.2144