Läs mer
5Alpha-Pregnane-3-one-20Alpha-ol, TRC
Produktkod. 30135076
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135076 100 mg
30005416 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30135076 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP705135100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 516-59-6
Molekylformel C21 H34 O2
Molekylvikt (g/mol) 318.49
Synonym Pregnan-3-one, 20-hydroxy-, (5α,20S)-, 5α-Pregnan-3-one, 20α-hydroxy- (6CI,7CI,8CI), (5α,20S)-20-Hydroxypregnan-3-one, 20α-Hydroxy-5α-pregan-3-one, 20α-Hydroxy-5α-pregnan-3-one, 5α,20α-Tetrahydroprogesterone, 5α-Pregnan-20α-ol-3-one, 5α-Pregnane-3-one-20α-ol, Allopregnan-20α-ol-3-one
IUPAC-namn (5S,8R,9S,10S,13S,14S,17S)-17-[(1S)-1-hydroxyethyl]-10,13-dimethyl-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-3-one
LEDER C[C@H](O)[C@H]1CC[C@H]2[C@@H]3CC[C@H]4CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifikationer
Kemiskt namn eller material 5alpha-Pregnane-3-one-20alpha-ol
Kvantitet 100 mg
InChI formel InChI=1S/C21H34O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h13-14,16-19,22H,4-12H2,1-3H3/t13-,14-,16-,17+,18-,19-,20-,21+/m0/s1
Formel vikt 318.2559