Läs mer
5Beta-Cholanic Acid-3Alpha,6Beta-diol, TRC
Produktkod. 30138000
Change view
Kvantitet:
2.5 mg
5 mg
10 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30138000 10 mg -
30132782 5 mg -
30007965 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138000 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC43143810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 668-49-5
Molekylformel C24H40O4
Molekylvikt (g/mol) 392.58
Synonym 3alpha,6beta-Dihydroxy-5beta-cholan-24-oic Acid, 3alpha,6beta-Dihydroxy-5beta-cholanic Acid, (3alpha,5beta,6beta)-3,6-Dihydroxycholan-24-oic Acid, 3alpha,6beta-Dihydroxy-5beta-cholan-24-oic Acid, 3alpha,6beta-Dihydroxy-5beta-cholanic Acid, 3alpha,6beta-Dihydroxy-5beta-cholanoic Acid, 3alpha,6beta-Dihydroxycholanoic Acid, 6beta-Hydroxylithocholic Acid, Murideoxycholic Acid, Murocholic Acid, NSC 18166;
IUPAC-namn (4R)-4-[(3R,5R,6R,8S,9S,10R,13R,14S,17R)-3,6-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
LEDER C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@@H]3C[C@@H](O)[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifikationer
Kemiskt namn eller material 5beta-Cholanic Acid-3alpha,6beta-diol
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C24H40O4/c1-14(4-7-22(27)28)17-5-6-18-16-13-21(26)20-12-15(25)8-10-24(20,3)19(16)9-11-23(17,18)2/h14-21,25-26H,4-13H2,1-3H3,(H,27,28)/t14-,15-,16+,17-,18+,19+,20+,21-,23-,24-/m1/s1
Formel vikt 392.2927
Procent renhet >95%