Läs mer
5Beta-Cholestan-3-one, TRC
Produktkod. 30368615
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30368615 250 mg
30367030 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30368615 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC431730250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 601-53-6
Molekylformel C27H46O
Molekylvikt (g/mol) 386.65
Synonym (5beta,17beta)-17-Octylandrostan-3-one, 3-Coprostanone, 5beta-Cholestanone, 5beta-Coprostan-3-one, Coprostanone, NSC 119031, beta-Cholestanone
IUPAC-namn (5R,8R,9S,10S,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-3-one
LEDER CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifikationer
Kemiskt namn eller material 5beta-Cholestan-3-one
Kvantitet 250 mg
InChI formel InChI=1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h18-20,22-25H,6-17H2,1-5H3/t19-,20-,22+,23-,24+,25+,26+,27-/m1/s1