Läs mer
5Beta-Dihydrocorticosterone, TRC
Produktkod. 17924485
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17924485 500 mg
17902175 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17924485 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD448785500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 566-01-8
Molekylformel C21H32O4
Molekylvikt (g/mol) 348.48
Synonym 11beta,21-Dihydroxy-5beta-pregnane-3,20-dione, (5R,8S,9S,10S,11S,13S,14S,17S)-11-Hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyltetradecahydro-1H-cyclopenta[a]phenanthren-3(2H)-one
LEDER C[C@]12CCC(=O)C[C@H]1CC[C@H]3[C@@H]4CC[C@H](C(=O)CO)[C@@]4(C)C[C@H](O)[C@H]23
Specifikationer
Kemiskt namn eller material 5beta-Dihydrocorticosterone
Kvantitet 500 mg
InChI formel InChI=1S/C21H32O4/c1-20-8-7-13(23)9-12(20)3-4-14-15-5-6-16(18(25)11-22)21(15,2)10-17(24)19(14)20/h12,14-17,19,22,24H,3-11H2,1-2H3/t12-,14+,15+,16-,17+,19-,20+,21+/m1/s1
Formel vikt 348.2301