Läs mer
5H-Dibenzo[a,d]cyclohepten-5-one (Dibenzosuberenone), Mikromol™
Produktkod. 30049697
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30049697 25 mg
17903246 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30049697 Leverantör Mikromol Leverantörsnummer MM0609.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
5H-Dibenzo[a,d]cyclohepten-5-one (Dibenzosuberenone)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5H-Dibenzo[a,d]cyclohepten-5-one (Dibenzosuberenone)
CAS 2222-33-5
Analyt- eller komponentnamn 5H-Dibenzo[a,d]cyclohepten-5-one (Dibenzosuberenone)
Frakt skick Room Temperature
Molekylformel C15 H10 O
Certifikat och efterlevnad ISO 17025
Synonym 5H-Dibenzo[a,d]cyclohepten-5-one, Dibenzosuberenone
InChI formel InChI=1S/C15H10O/c16-15-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)15/h1-10H
LEDER O=C1c2ccccc2C=Cc3ccccc13
IUPAC-namn dibenzo[2,1-b:1',2'-e][7]annulen-11-one
Molekylvikt (g/mol) 206.24
Kvantitet 25 mg
Formel vikt 206.0732 g/mol