Läs mer
5H-Pyrido[3,2-b]azepine-6,9-(7H,8H)-dione, TRC
Produktkod. 30422992
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30422992 25 mg
30425413 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30422992 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP99171025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 676596-63-7
Molekylformel C9 H8 N2 O2
Molekylvikt (g/mol) 176.17
Synonym 5H-Pyrido[3,2-b]azepine-6,9-dione, 7,8-dihydro-, 5H-Pyrido[3,2-b]azepine-6,9(7H,8H)-dione, 7,8-Dihydro-5H-pyrido[3,2-b]azepine-6,9-dione
IUPAC-namn 7,8-dihydro-5H-pyrido[3,2-b]azepine-6,9-dione
LEDER O=C1CCC(=O)c2ncccc2N1
Specifikationer
Kemiskt namn eller material 5H-Pyrido[3,2-b]azepine-6,9-(7H,8H)-dione
Kvantitet 25 mg
InChI formel InChI=1S/C9H8N2O2/c12-7-3-4-8(13)11-6-2-1-5-10-9(6)7/h1-2,5H,3-4H2,(H,11,13)
Formel vikt 176.0586