Läs mer
Tocris Bioscience™ 5J 4
CRAC channel blocker; anti-inflammatory
Brand: Tocris Bioscience™ 5490/50
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
5J 4 is a calcium release-activated calcium (CRAC) channel blocker. Blocks store-operated calcium entry (SOCE) in HeLa-O+S cells and Th17 cells in vitro. Ameliorates symptoms and reduces CNS infiltration by mononuclear cells in a mouse model of experimental autoimmune encephalomyelitis (EAE) in vivo.Specifikationer
| N-[[(6-Hydroxy-1-naphthalenyl)amino]thioxomethyl]-2-furancarboxamide | |
| 827001-82-1 | |
| 50 mg |