Läs mer
(5R)-Tetrahydro-5-methyl-2H-pyran-2-one, TRC
Produktkod. 30417184
Change view
Kvantitet:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30417184 500 mg
30428617 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30417184 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT293970500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2857-75-2
Molekylformel C6H10O2
Molekylvikt (g/mol) 114.14
Synonym (R)-Tetrahydro-5-methyl-2H-pyran-2-one
IUPAC-namn (5R)-5-methyloxan-2-one
LEDER C[C@@H]1CCC(=O)OC1
Specifications
Kemiskt namn eller material (5R)-Tetrahydro-5-methyl-2H-pyran-2-one
Kvantitet 500 mg
InChI formel InChI=1S/C6H10O2/c1-5-2-3-6(7)8-4-5/h5H,2-4H2,1H3/t5-/m1/s1
Formel vikt 114.0681