Läs mer
(5RS)-5-(4-Methoxyphenyl)-3-methyl-1-oxa-3-azaspiro[5.5]undecane Hydrochloride, Mikromol™
Produktkod. 30460865
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30460865 4 x 25 mg
17933366 25 mg
17953766 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30460865 Leverantör Mikromol Leverantörsnummer MM0393.0900254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(5RS)-5-(4-Methoxyphenyl)-3-methyl-1-oxa-3-azaspiro[5.5]undecane Hydrochloride, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (5RS)-5-(4-Methoxyphenyl)-3-methyl-1-oxa-3-azaspiro[5.5]undecane Hydrochloride
CAS 93413-56-0
Analyt- eller komponentnamn (5RS)-5-(4-Methoxyphenyl)-3-methyl-1-oxa-3-azaspiro[5.5]undecane Hydrochloride
Frakt skick Room Temperature
Molekylformel C17 H25 N O2 . Cl H
Certifikat och efterlevnad ISO 17025
Synonym Venlafaxine Hydrochloride Imp. E (EP), Venlafaxine Imp. E (EP), (5RS)-5-(4-Methoxyphenyl)-3-methyl-1-oxa-3-azaspiro[5.5]undecane Hydrochloride, Venlafaxine Hydrochloride Imp. E (EP) as Hydrochloride, Venlafaxine Hydrochloride Impurity E as Hydrochloride, Venlafaxine Impurity E as Hydrochloride
InChI formel InChI=1S/C17H25NO2.ClH/c1-18-12-16(14-6-8-15(19-2)9-7-14)17(20-13-18)10-4-3-5-11-17;/h6-9,16H,3-5,10-13H2,1-2H3;1H
Lösningstyp Neat
LEDER Cl.COc1ccc(cc1)C2CN(C)COC23CCCCC3
IUPAC-namn 5-(4-methoxyphenyl)-3-methyl-1-oxa-3-azaspiro[5.5]undecane;hydrochloride
Molekylvikt (g/mol) 311.85
Kvantitet 4 x 25 mg
Formel vikt 311.1652 g/mol