Läs mer
6-(1,3-Dioxoisoindolin-2-yl)hexanoic Acid, TRC
Produktkod. 30369638
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30369638 10 g
1 options
Produktkod. 30369638 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD48679510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6-(1,3-Dioxoisoindolin-2-yl)hexanoic Acid
CAS 4443-26-9
Molekylformel C14 H15 N O4
Kvantitet 10 g
Synonym 1,3-Dioxo-2-isoindolinehexanoic Acid, 1,3-Dihydro-1,3-dioxo-2H-isoindole-2-hexanoic Acid, 6-(1,3-Dioxo-1,3-dihydroisoindol-2-yl)hexanoic Acid, 6-Phthalimidohexanoic Acid, Phthalimidocapronic Acid, epsilon-Phthalimidocaproic Acid, ω-Phthalimidocaproic Acid
InChI formel InChI=1S/C14H15NO4/c16-12(17)8-2-1-5-9-15-13(18)10-6-3-4-7-11(10)14(15)19/h3-4,6-7H,1-2,5,8-9H2,(H,16,17)
LEDER O=C(O)CCCCCN1C(=O)C=2C=CC=CC2C1=O
IUPAC-namn 6-(1,3-dioxoisoindolin-2-yl)hexanoic acid
Molekylvikt (g/mol) 261.27
Formel vikt 261.1001