Läs mer
6-(1-Fluoroethyl)-1,3,5-triazine-2,4-diamine, TRC
Produktkod. 30180214
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180214 250 mg
30179857 500 mg
30179816 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30180214 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF532080250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1637285-20-1
Molekylformel C5 H8 F N5
Molekylvikt (g/mol) 157.15
Synonym 1,3,5-Triazine-2,4-diamine, 6-(1-fluoroethyl)-
IUPAC-namn 6-(1-fluoroethyl)-1,3,5-triazine-2,4-diamine
LEDER CC(F)c1nc(N)nc(N)n1
Specifikationer
Kemiskt namn eller material Indaziflam-desindenyl
Kvantitet 250 mg
InChI formel InChI=1S/C5H8FN5/c1-2(6)3-9-4(7)11-5(8)10-3/h2H,1H3,(H4,7,8,9,10,11)
Formel vikt 157.0764