Läs mer
6-[(3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydronaphthalen-2-yl)carbonyl] Nicotinic Acid Methyl Ester, TRC
Produktkod. 30426069
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30426069 250 mg
1 options
Produktkod. 30426069 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP271770250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6-[(3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydronaphthalen-2-yl)carbonyl] Nicotinic Acid Methyl Ester
CAS 153559-92-3
Molekylformel C23H27NO3
Kvantitet 250 mg
Synonym 6-[(5,6,7,8-Tetrahydro-3,5,5,8,8-pentamethyl-2-naphthalenyl)carbonyl]-3-pyridinecarboxylic Acid Methyl Ester
InChI formel InChI=1S/C23H27NO3/c1-14-11-17-18(23(4,5)10-9-22(17,2)3)12-16(14)20(25)19-8-7-15(13-24-19)21(26)27-6/h7-8,11-13H,9-10H2,1-6H3
LEDER COC(=O)c1ccc(nc1)C(=O)c2cc3c(cc2C)C(C)(C)CCC3(C)C
IUPAC-namn methyl 6-(3,5,5,8,8-pentamethyl-6,7-dihydronaphthalene-2-carbonyl)pyridine-3-carboxylate
Molekylvikt (g/mol) 365.47
Formel vikt 365.1991