Läs mer
6-[(3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethenyl] Nicotinic Acid Methyl Ester, TRC
Produktkod. 30427769
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30427769 25 mg
1 options
Produktkod. 30427769 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP27178025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6-[(3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethenyl] Nicotinic Acid Methyl Ester
CAS 153559-44-5
Molekylformel C24H29NO2
Kvantitet 25 mg
Synonym 6-[1-(5,6,7,8-Tetrahydro-3,5,5,8,8-pentamethyl-2-naphthalenyl)ethenyl]-3-pyridinecarboxylic Acid Methyl Ester
InChI formel InChI=1S/C24H29NO2/c1-15-12-19-20(24(5,6)11-10-23(19,3)4)13-18(15)16(2)21-9-8-17(14-25-21)22(26)27-7/h8-9,12-14H,2,10-11H2,1,3-7H3
LEDER COC(=O)c1ccc(nc1)C(=C)c2cc3c(cc2C)C(C)(C)CCC3(C)C
IUPAC-namn methyl 6-[1-(3,5,5,8,8-pentamethyl-6,7-dihydronaphthalen-2-yl)ethenyl]pyridine-3-carboxylate
Molekylvikt (g/mol) 363.49
Formel vikt 363.2198