Läs mer
6-[4-(1-Cyclohexyl-1H-tetrazol-5-yl)butoxy]-1-[4-(1-cyclohexyl-1H-tetrazol-5-yl)butyl]-3,4-dihydro-2(1H)-quinolinone, TRC
Produktkod. 30179902
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179902 10 mg
30180027 100 mg
30180316 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179902 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC98825510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 865792-18-3
Molekylformel C31 H45 N9 O2
Molekylvikt (g/mol) 575.75
Synonym 2(1H)-Quinolinone, 6-[4-(1-cyclohexyl-1H-tetrazol-5-yl)butoxy]-1-[4-(1-cyclohexyl-1H-tetrazol-5-yl)butyl]-3,4-dihydro-, 6-[4-(1-Cyclohexyl-1H-tetrazol-5-yl)butoxy]-1-[4-(1-cyclohexyl-1H-tetrazol-5-yl)butyl]-3,4-dihydro-2(1H)-quinolinone, 1-(4-(5-Cyclohexyl-1H-tetrazol-1-yl)butyl)-6-(4-(1-cyclohexyl-1H-tetrazol-5-yl)butoxy)-3,4-dihydroquinolin-2(1H)-one
IUPAC-namn 6-[4-(1-cyclohexyltetrazol-5-yl)butoxy]-1-[4-(1-cyclohexyltetrazol-5-yl)butyl]-3,4-dihydroquinolin-2-one
LEDER O=C1CCc2cc(OCCCCc3nnnn3C4CCCCC4)ccc2N1CCCCc5nnnn5C6CCCCC6
Specifikationer
Kemiskt namn eller material 1-(4-(5-Cyclohexyl-1H-tetrazol-1-yl)butyl)-6-(4-(1-cyclohexyl-1H-tetrazol-5-yl)butoxy)-3,4-dihydroquinolin-2(1H)-one
Kvantitet 10 mg
InChI formel InChI=1S/C31H45N9O2/c41-31-20-17-24-23-27(42-22-10-8-16-30-33-35-37-40(30)26-13-5-2-6-14-26)18-19-28(24)38(31)21-9-7-15-29-32-34-36-39(29)25-11-3-1-4-12-25/h18-19,23,25-26H,1-17,20-22H2
Formel vikt 575.3696