Läs mer
6-[4-(3-Trifluoromethylpyridin-2-yloxy)-2-methylphenyl]-1,5-dimethyl-2,4(1H,3H)-pyrimidinedione, TRC
Produktkod. 30454348
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30454348 5 mg
1 missing translation for 'options'
Product Code. 30454348 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCT7915505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material 6-[4-(3-Trifluoromethylpyridin-2-yloxy)-2-methylphenyl]-1,5-dimethyl-2,4(1H,3H)-pyrimidinedione
CAS 1643462-64-9
Molekylformel C19 H16 F3 N3 O3
Kvantitet 5 mg
Synonym 1,5-Dimethyl-6-(2-methyl-4-((3-(trifluoromethyl)pyridin-2-yl)oxy)phenyl)pyrimidine-2,4(1H,3H)-dione
InChI formel InChI=1S/C19H16F3N3O3/c1-10-9-12(28-17-14(19(20,21)22)5-4-8-23-17)6-7-13(10)15-11(2)16(26)24-18(27)25(15)3/h4-9H,1-3H3,(H,24,26,27)
LEDER CN1C(=O)NC(=O)C(=C1c2ccc(Oc3ncccc3C(F)(F)F)cc2C)C
IUPAC-namn 1,5-dimethyl-6-[2-methyl-4-[[3-(trifluoromethyl)-2-pyridyl]oxy]phenyl]pyrimidine-2,4-dione
Molekylvikt (g/mol) 391.34
Formel vikt 391.1144