Läs mer
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isoquinoline (>85%), TRC
Produktkod. 30410197
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30410197 50 mg
30423951 500 mg
30423179 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30410197 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT30325550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 675576-26-8
Molekylformel C15H18BNO2
Molekylvikt (g/mol) 255.11
IUPAC-namn 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isoquinoline
LEDER CC1(C)OB(OC1(C)C)c2ccc3cnccc3c2
Specifikationer
Kemiskt namn eller material 6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isoquinoline
Kvantitet 50 mg
InChI formel InChI=1S/C15H18BNO2/c1-14(2)15(3,4)19-16(18-14)13-6-5-12-10-17-8-7-11(12)9-13/h5-10H,1-4H3
Formel vikt 255.1431