missing translation for 'onlineSavingsMsg'
Learn More

6-(4-Bromophenyl)imidazo[2,1-b]thiazole, Alfa Aesar™

948.70 SEK - 3406.41 SEK

Chemical Identifiers

CAS 7120-13-0
Molecular Formula C11H7BrN2S
Molecular Weight (g/mol) 279.155
MDL Number MFCD00269210
InChI Key GIARQEDFNKBIQY-UHFFFAOYSA-N
Synonym 6-4-bromophenyl imidazo 2,1-b thiazole, 6-4-bromophenyl imidazo 2,1-b 1,3 thiazole, 6-4-bromo-phenyl-imidazo 2,1-b thiazole, acmc-20am12, 6-4-bromophenyl imidazo 2,1-b-1,3-thiazole, 6-4-bromophenyl imidazo 2,1-b 1,3-thiazoline
PubChem CID 667712
IUPAC Name 6-(4-bromophenyl)imidazo[2,1-b][1,3]thiazole
SMILES C1=CC(=CC=C1C2=CN3C=CSC3=N2)Br
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11490040 SDS View Documents Alfa Aesar
H50572.MD
250mg 948.70 SEK
250mg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 21-10-2019
Add to basket
 
11400050 SDS View Documents Alfa Aesar
H50572.03
1g 3406.41 SEK
1g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 21-10-2019
Add to basket
 
Specifications

Chemical Identifiers

7120-13-0
279.155
GIARQEDFNKBIQY-UHFFFAOYSA-N
667712
C1=CC(=CC=C1C2=CN3C=CSC3=N2)Br
C11H7BrN2S
MFCD00269210
6-4-bromophenyl imidazo 2,1-b thiazole, 6-4-bromophenyl imidazo 2,1-b 1,3 thiazole, 6-4-bromo-phenyl-imidazo 2,1-b thiazole, acmc-20am12, 6-4-bromophenyl imidazo 2,1-b-1,3-thiazole, 6-4-bromophenyl imidazo 2,1-b 1,3-thiazoline
6-(4-bromophenyl)imidazo[2,1-b][1,3]thiazole

Specifications

250mg
7120-13-0
MFCD00269210
GIARQEDFNKBIQY-UHFFFAOYSA-N
6-(4-bromophenyl)imidazo[2,1-b][1,3]thiazole
667712
6-(4-Bromophenyl)imidazo[2,1-b]thiazole
C11H7BrN2S
6-4-bromophenyl imidazo 2,1-b thiazole, 6-4-bromophenyl imidazo 2,1-b 1,3 thiazole, 6-4-bromo-phenyl-imidazo 2,1-b thiazole, acmc-20am12, 6-4-bromophenyl imidazo 2,1-b-1,3-thiazole, 6-4-bromophenyl imidazo 2,1-b 1,3-thiazoline
C1=CC(=CC=C1C2=CN3C=CSC3=N2)Br
279.155
279.16