Läs mer
6-(4-Phenylbutoxy)-1-hexanamine, TRC
Produktkod. 17912215
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17912215 250 mg
17910765 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17912215 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP297920250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 97664-58-9
Molekylformel C16 H27 N O
Molekylvikt (g/mol) 249.39
Synonym D 2543
IUPAC-namn 6-(4-phenylbutoxy)hexan-1-amine
LEDER NCCCCCCOCCCCc1ccccc1
Specifikationer
Kemiskt namn eller material 6-(4-Phenylbutoxy)-1-hexanamine
Kvantitet 250 mg
InChI formel InChI=1S/C16H27NO/c17-13-7-1-2-8-14-18-15-9-6-12-16-10-4-3-5-11-16/h3-5,10-11H,1-2,6-9,12-15,17H2
Formel vikt 249.2093